Learn More
anti-CD8, Alexa Fluor 750, Clone: 53-6.7, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB116S100
 View more versions of this product
Catalog No. FAB116S100
Add to cart
Specifications
Specifications
CD8
53-6.7
Flow Cytometry 5 uL/10^6 cells
Supernatant
CD8A
Mouse thymus or spleen
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
925
Mouse
Flow Cytometry
Alexa Fluor 750
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD8, CD8A, CD8a molecule, LEU2, p32
Rat
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects both the alpha and alpha ' chains of mouse CD8 (1).