Learn More
CD4/CD8 Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112269
Catalog No. 01-670-964
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils. CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells.
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD4 (clone 7E14, isotype IgG1).
Specifications
Specifications
CD4/CD8
Cocktail
FITC, PE
CD8A
Liquid
CD4, CD8A, CD8B
human peripheral blood T lymphocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
17D8
PBS with 0.2% carrier protein and 0.08% sodium azide
P01730, P01732, P10966
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4, CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
920, 925, 926