Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD8, Clone: 53-6.7, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB116V025
This item is not returnable.
View return policy
Specifications
CD8 | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD8A | |
Mouse thymus or spleen | |
25 Tests | |
Primary | |
Detects both the alpha and alpha ' chains of mouse CD8 (1). | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Flow Cytometry | |
53-6.7 | |
Flow Cytometry 5 uL/10^6 cells | |
CD8, CD8A, CD8a molecule, LEU2, p32 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
925 | |
Mouse | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction