Learn More
anti-CD8, Clone: 53-6.7, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB116V100
 View more versions of this product
Catalog No. FAB116V100
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
CD8
53-6.7
Flow Cytometry 5 uL/10^6 cells
Supernatant
CD8A
Mouse thymus or spleen
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
925
Mouse
Flow Cytometry
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD8, CD8A, CD8a molecule, LEU2, p32
Rat
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects both the alpha and alpha ' chains of mouse CD8 (1).