Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD8, Clone: 53-6.7, R&D Systems™

Rat Monoclonal Antibody

Specifications

Antigen CD8
Immunogen Mouse thymus or spleen
Clone 53-6.7
Applications Flow Cytometry
Conjugate Alexa Fluor 405
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FAB116V025
View Documents
R&D Systems
FAB116V025
25 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FAB116V100
View Documents
R&D Systems
FAB116V100
100 Tests This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

CD8
53-6.7
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Monoclonal
Primary
Detects both the alpha and alpha ' chains of mouse CD8 (1).
Protein A or G purified from hybridoma culture supernatant
Mouse thymus or spleen
Flow Cytometry
Supernatant
Rat
IgG2a
925
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit