Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Mouse anti-Human, SPRD (R-phycoerythrin/Cyanine 5), Clone: RFT8, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 953613
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD8 | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Thymocytes and E-rosetted lymphocytes | |
RUO | |
Human | |
IgG1 |
Flow Cytometry | |
RFT8 | |
SPRD | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD8 Mouse anti-Human, SPRD (R-phycoerythrin/Cyanine 5), Clone: RFT8, Southern Biotech™ > R-phycoerythrin/Cyanine 5, 100 tests
Spot an opportunity for improvement?Share a Content Correction