Learn More
CD8/HLA-DR Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112174
Catalog No. 01-670-893
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated HLADR (clone 423L, isotype IgG2a).
CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells.
Specifications
Specifications
CD8/HLA-DR
Cocktail
FITC, PE
CD74
Liquid
CD74, CD8A, CD8B
human peripheral blood T lymphocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
17D8, 423L
PBS with 0.2% carrier protein and 0.08% sodium azide
P01732, P04233, P10966
CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta, CD74 antigen, HLA class II histocompatibility antigen gamma chain, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia-associated invariant chain, MHC HLA-DR gamma chain, gamma chain of class II antigens, ia antigen-associated invariant chain, HLADG, DHLAG, IA-gamma
Mouse
IgG1/IgG2a
50 Tests
4° C, do not freeze
Primary
925, 926, 972