Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Mouse anti-Human, Clone: AS29, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112159

Catalog No. 01-670-878

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody recognizes the extra-cellular domain of human CD80; it does not bind the extracellular domain of CD86.

Involved in the costimulatory signal essential for T lymphocytes activation. T cell proliferation and cytokine production is induced by the binding of CD28 or CTLA4 to this receptor.
Specifications

Specifications

CD80
Monoclonal
0.6 mg/mL
PBS with 0.2% carrier protein and 0.08% sodium azide
P33681
B7-1, B7.1, Cd28l, Ly-53, Ly53, MIC17, TS/A-1, LAB7, B7.1, CD28LG, CD28LG1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
941
Immunoprecipitation, Neutralization
AS29
Unconjugated
CD80
Liquid
CD80
Recombit extra-cellular domain of human CD80
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit