Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD81 Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems MAB4865
This item is not returnable.
View return policy
Description
CD81 Monoclonal specifically detects CD81 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD81 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_598416 | |
CD81 | |
NS0 mouse myeloma cell line transfected with mouse CD81 Met1-Tyr236 Accession # NP_598416 | |
100 μg | |
Primary | |
Detects mouse CD81. Stains mouse CD81 transfectants but not irrelevant transfectants. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
431301 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD81, CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, TSPAN28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
975 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction