Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 Mouse anti-Human, Clone: B72-H2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112172

Catalog No. 01-670-891

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD86 is one of two ligands (the other CD80) for CTLA4 and CD28. CD86 acts as costimulatory molecule in eliciting T-cell help during antigen presentation. Antigen presentation in the absence of sufficient costimulation involving CD86/CD80 can induce tolerance. CD80 appears to play a role distinct from CD80 in T helper cell differentiation.

This antibody recognizes the extra-cellular domain of human CD86; it does not bind the extracellular domain of CD80.
Specifications

Specifications

CD86
Monoclonal
0.375 mg/mL
Assay Dependent
CD86
Liquid
CD86
Recombit extracellular domain of human CD86.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry
B72-H2
Unconjugated
PBS with 0.08% sodium azide
P42081
B7-2, B7.2, B70, CD28LG2, LAB72, MGC34413, B-lymphocyte activation antigen B7-2, B-lymphocyte antigen B7-2, B7-2 antigen, BU63, CD28 antigen ligand 2
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
942
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit