Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD86 Mouse anti-Human, Clone: IT2.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041528
Description
Anti-CD86 Antibody, clone IT2.2 detects level of CD86 and has been published and validated for use in flow cytometry, western blot, immunohistochemistry.Specifications
CD86 | |
Monoclonal | |
Unconjugated | |
NP_008820 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Flow Cytometry, Immunohistochemistry, Western Blot | |
IT2.2 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
CD86, B7-2, B70, BU63, CD28LG2, FUN-1, LAB72 | |
Human LCL whole cells. | |
50 μL | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only