Learn More
CD86 Mouse anti-Human, V450, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
$205.50 - $447.50
Specifications
| Antigen | CD86 |
|---|---|
| Clone | 2331 (FUN-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Violet Fluor 450 (V450) |
Description
2331 (FUN-1) recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Mfr. No. 555663) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: 2331(FUN-1)
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD86 | |
| Flow Cytometry | |
| Violet Fluor 450 (V450) | |
| RUO | |
| Human | |
| Human HBL-1 Cell Line | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 2331 (FUN-1) | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.