Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCAR (soluble) Mouse anti-Human, FITC, Clone: MIP8a, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135974

Catalog No. 01672459


Add to cart

Description

Description

Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Specifications

Specifications

FCAR (soluble)
Monoclonal
0.1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
P24071
CD89, Fc alpha receptor, IgA Fc receptor, immunoglobulin alpha Fc receptor
Mouse
IgG1
100 μg
4°C or -20°C if preferred
Primary
2204
Flow Cytometry
MIP8a
FITC
FCAR
Liquid
FCAR
Recombit soluble human Fc alpha R.
Purified
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit