Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD8a APC, Clone: 53-6.7,
Rat Monoclonal Antibody
$184.00 - $273.00
Specifications
Antigen | CD8a |
---|---|
Clone | 53-6.7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1535MI
|
MilliporeSigma™
MABF1535 |
25 μg |
Each for $184.00
|
|
|||||
MABF1536MI
|
MilliporeSigma™
MABF1536 |
100 μg |
Each for $273.00
|
|
|||||
Description
Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry
The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.Specifications
CD8a | |
Flow Cytometry | |
APC | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
CD8a | |
IgG2a κ | |
Purified by size-exclusion chromatography |
53-6.7 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P01731 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title