Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD8a FITC, Clone: 53-6.7,

Rat Monoclonal Antibody

$93.70 - $153.50

Specifications

Antigen CD8a
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1541MI
View Documents
MilliporeSigma™
MABF1541
25μg
Each for $153.50
Add to cart
 
MABF1542MI
View Documents
MilliporeSigma™
MABF1542
100μg
Each for $93.70
Add to cart
 
Description

Description

Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry

The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.
Specifications

Specifications

CD8a
Monoclonal
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
IgG2a, κ
P01731
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
Primary
Purified
53-6.7
Rat
Proprietary immunogen
CD8a
RUO
Purified by size-exclusion chromatography
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit