Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 beta Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511496

Catalog No. PIPA511496


Add to cart

Description

Description

The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigen displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains.
Specifications

Specifications

CD8 beta
Polyclonal
PBS with 0.09% sodium azide
P10966
Cd8b1, CD8 antigen 37 kDa chain, CD8 antigen beta-chain, CD8 antigen beta chain, OX-8 membrane antigen, T-cell surface glycoprotein CD8 beta chain
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
926
Flow Cytometry, Western Blot
Unconjugated
CD8B
Liquid
CD8B
KLH conjugated synthetic peptide between 64-92 amino acids from the N-terminal region of human CD8B
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit