Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Rat anti-Mouse, Brilliant Violet 421, Clone: KMC8, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564235

Catalog No. BDB564235


Add to cart

Description

Description

The KMC8 monoclonal antibody specifically binds to CD9 which is also known as Tetraspanin-29 (Tspan29). CD9 is a 24-kDa member of the transmembrane 4 superfamily, also called the tretraspanin family. In the mouse, CD9 is present on bone marrow myeloid cells, stromal cells, and megakaryocyte-committed progenitors; subsets of peripheral T and B lymphocytes; and neutrophils, platelets, dendritic cells, and bone marrow-derived macrophages. CD9 has been found to be associated with integrins and other cell-surface receptors. It appears to play roles in signal transduction and in regulating cellular adhesive properties. CD9 can also reportedly participate in T-cell costimulation and induction of apoptosis. The KMC8 antibody reportedly blocks certain CD9 functions and activates macrophages.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CD9
KMC8
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Cd9; Tetraspanin-29; Tspan29
Affinity Purified
(C57BL/6 x DBA/2)F1 mouse bone marrow-derived stromal cell line BMS2
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit