Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, Brilliant Violet 510, Clone: M-L13, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563640

Catalog No. BDB563640


Add to cart

Description

Description

The M-L13 monoclonal antibody specifically binds to a 24 kDa type III transmembrane protein that is expressed on platelets, pre-B cells, monocytes, endothelial and epithelial cells. CD9 belongs to a family of membrane proteins called tetraspanins that transverse the membrane four times. CD9 is weakly expressed on resting mature B cells. M-L13 induces platelet aggregation and activation. This antibody is also suitable for staining acetone-fixed, frozen tissue sections.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications

Specifications

CD9
M-L13
CD9 antigen (p24); 5H9; BA2; BTCC-1; DRAP-27; GIG2; MIC3; MRP-1; TSPAN29
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit