Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD90.2, Violer Fluor 450, Clone: 30-H12,

Rat Monoclonal Antibody

$153.50 - $272.50

Specifications

Antigen CD90.2
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1450MI
View Documents
MilliporeSigma™
MABF1450
25μg
Each for $153.50
Add to cart
 
MABF1451MI
View Documents
MilliporeSigma™
MABF1451
100μg
Each for $272.50
Add to cart
 
Description

Description

Specifically detects CD90.2 Clone: 30-H12 in Mouse samples, and it is validated for Flow Cytometry

The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.
Specifications

Specifications

CD90.2
Monoclonal
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
IgG2b
P01831
2°C to 8°C for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
Primary
Purified
30-H12
Rat
Proprietary immunogen
Thy1, Thy-1
RUO
Purified by size-exclusion chromatography
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit