Learn More
Human CD90/Thy1 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
Supplier:  R&D Systems FAB2067A100
 View more versions of this product
Catalog No. FAB2067A100
Add to Cart
This item is not returnable. View return policy
Description
Description
CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD90/Thy1
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7070
Human
Supernatant
Flow Cytometry
Thy-1A1
Flow Cytometry 10 uL/10^6 cells
P04216
THY1
Human CD90/Thy1 Accession # P04216
100 Tests
Primary
Detects human CD90/Thy1.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a
Product Suggestions
Product Suggestions