Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD93 Mouse anti-Human, APC, Clone: 273107, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523615

Catalog No. PIMA523615

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse C1q R1 is observed.

The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.
Specifications

Specifications

CD93
Monoclonal
APC
CD93
Liquid
CD93
Mouse myeloma cell line NS0-derived recombit human C1qR1/CD93 Ala24-Lys580
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
273107
Buffered saline solution with BSA and 0.09% sodium azide
Q9NPY3
C1QR1, C1qRP, CDw93, ECSM3, MXRA4, dJ737E23.1, C1qR, C1q receptor
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
22918
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit