Learn More
CD96 Mouse anti-Human, BB515, Clone: 6F9, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564774
Catalog No. BDB564774
Add to cart
Description
Description
The 6F9 monoclonal antibody specifically binds to human CD96, also known as TACTILE (T cell activation increased late expression). CD96 is a type I transmembrane glycoprotein and member of the Ig superfamily. CD96 is expressed at low levels on resting natural killer (NK) cells and T cells and at high levels on activated NK and T cells. CD96 is also expressed on some T-cell leukemia and acute myeloid leukemia cells. CD96 may serve as a marker for acute myelogenous leukemia stem cells. CD96 plays a role in the adhesive interactions of activated NK and T cells during immune responses. CD96 binds to the poliovirus receptor (CD155) that is highly expressed by some tumor cells. CD155-mediated ligation of CD96 can induce NK cell-mediated cytotoxicity. CD96-mediated uptake of CD155 may adversely affect NK cells and thus reduce their effectiveness in anti-tumor responses.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Specifications
Specifications
CD96
6F9
TACTILE; T cell activation increased late expression
Affinity purified
Human CD96 Transfected Cell Line
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide
Mouse
IgG1, κ
RUO
Primary
Human