Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD96 v2 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6199A
This item is not returnable.
View return policy
Description
CD96 Monoclonal specifically detects CD96 in Human samples. It is validated for Flow Cytometry.Specifications
CD96 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD96, CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILE, TACTILEincreased late expression, T-cell surface protein tactile | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10225 | |
Human | |
Purified |
Flow Cytometry | |
628211 | |
Flow Cytometry | |
NP_005807 | |
CD96 | |
Mouse myeloma cell line NS0-derived recombinant human CD96 Val22-Asp501 Accession # NP_005807 | |
100 Tests | |
Primary | |
Detects human CD96 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction