Learn More
anti-CD97, Alexa Fluor 488, Clone: 587702, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB3734G
Catalog No. FAB3734G
Add to cart
This item is not returnable. View return policy
Description
Description
CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD97 Isoform 1
587702
Flow Cytometry 5 uL/10^6 cells
AAH06676
CD97
Chinese hamster ovary cell line CHO-derived recombinant mouse CD97v2 Gln24-His384 Accession # AAH06676
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
976
Mouse
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
Rat
IgG1
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) CD97 and no cross-reactivity with recombinant human (rh) CD96v2, rhCD97, or rmCD96 is observed.