Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528350
Description
Recommended positive controls: A549, HeLa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CD97 Polyclonal antibody specifically detects CD97 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD97 | |
Polyclonal | |
Unconjugated | |
ADGRE5 | |
AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
976 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P48960 | |
ADGRE5, CD97 | |
Recombinant fragment corresponding to a region within amino acids 320 and 751 of Human CD97 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction