Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512498
Description
CDA forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. This protein is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in the CDA gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.Specifications
CDA | |
Polyclonal | |
CDA | |
Cytidine deaminase; Cytidine aminohydrolase; CDD | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
72269, 978 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P32320, P56389 | |
CDA | |
KLH conjugated synthetic peptide between 51-83 amino acids from the central region of human CDA | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title