Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDC2/CDK1 (p Thr161), Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP14177050UG

Catalog No. NBP14177050

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CDC2/CDK1 (p Thr161)
1 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100
Affinity Purified
CDK1
synthesized phosphopeptide derived from human CDC2 around the phosphorylation site of threonine161 (T-Y-TP-H-E).
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
CDC28A, CDC2MGC111195, cell cycle controller CDC2, Cell division control protein 2 homolog, Cell division protein kinase 1, cyclin-dependent kinase 1, DKFZp686L20222, EC 2.7.11.22, EC 2.7.11.23, G1 to S and G2 to M, p34 protein kinase, P34CDC2
Rabbit
IgG
50 ug
Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primary
983
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only