Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cdc20 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526145
Description
This antibody is predicted to react with hamster, mouse and rat based on sequence homology.
CDC20 is an activating regulatory factor for the APC/C (anaphase promoting complex/cyclosome). It activates the ubiquitination activity of the APC/C. CDC20 confers a strict destruction-box (D-box) dependence on APC. Levels of CDC20, as well as its binding to APC, peak in mitosis and decrease drastically at early G1.Specifications
CDC20 | |
Polyclonal | |
Unconjugated | |
Q12834 | |
CDC20 | |
KLH conjugated synthetic peptide between 57-84 amino acids from the N-terminal region of human CDC20 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
CDC20 | |
CDC20A, bA276H19.3, p55CDC, CDC20 cell division cycle 20 homolog, cell division cycle protein 20 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
991 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction