Learn More
Cdc25A Mouse anti-Human, Rat, Clone: DCS120, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05743
Catalog No. 05-743-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-Cdc25A Antibody, clone DCS120 is a high quality Mouse Monoclonal Antibody for the detection of Cdc25A and has been validated in immunoprecipitation, western blotting and immunohistochemistry.
Specifications
Specifications
Cdc25A
DCS120
10mM PBS, pH 7.4
Purified
Murine
IgG2a
100μg
Primary
Human, Rat
Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_001789
CDC25A, CDC25A2
Purified recombinant cdc25A
Protein A purfied
−20°C for 24 months.
Monoclonal