Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDC25C Ab-7, Clone: 25C07 (TC-113), Epredia™ LabVision

Mouse Monoclonal Antibody

Specifications

Antigen CDC25C Ab-7
Classification Monoclonal
Host Species Mouse
Target Species Human
Isotype IgG1 κ
View More Specs

Products 7
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate Concentration Quantity Price Quantity & Availability  
MS1187B0
View Documents
Epredia
MS1187B0
Biotin 200μg/mL 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187B
View Documents
Epredia
MS1187B
Biotin 200μg/mL 1 mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187P1
View Documents
Epredia
MS1187P1
Unconjugated 200μg/mL 500 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187P1ABX
View Documents
Epredia
MS1187P1ABX
Unconjugated 1mg/mL 100 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187PABX
View Documents
Epredia
MS1187PABX
Unconjugated 1mg/mL 200 μL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187R7
View Documents
Epredia
MS1187R7
Unconjugated 7 mL Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
MS1187PCS
View Documents
Epredia
MS1187PCS
Unconjugated 5 Slides Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Cyclin-dependent kinases can be activated by CDC25, which removes inhibitory phosphates from tyrosine and threonine residues. At least three CDC25 genes (CDC25A, CDC25B, and CDC25C) have been identified in humans. Whereas CDC25A and CDC25B are expressed throughout the cell cycle, with peak expression in G1 for CDC25A and in both G1-S-phase and G2 for CDC25B, CDC25C is predominantly expressed in G2.

Host Species: Mouse
Clone: 25C07 (TC-113)
Isotype: IgG1/κ
Cross Reactivity: Human. Does not react with mouse and rat. Others-not tested.
Epitope: aa222-246
Immunogen: Recombinant human CDC25C protein.
Molecular Weight: 53-56kDa
Positive Control: HeLa, HT29, or LS174T cells. Breast CA.
Cellular Localization: Cytoplasmic

Ab-7 is excellent for the staining of formalin-fixed, paraffin-embedded tissues.

Immunofluorescence, Immunohistochemistry (Formalin/Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

CDC25C Ab-7
Mouse
IgG1 κ
Monoclonal
Human
Primary
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit