Learn More
Cdc25C Mouse anti-Human, Mouse, Clone: TC-15, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05507
Catalog No. 05-507-MI
Add to cart
Description
Description
Detect Cdc25C with Anti-Cdc25C Antibody, clone TC-15 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
Specifications
Specifications
Cdc25C
TC-15
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
100μg
Primary
Human, Murine
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_001790
CDC25C, CDC25
GST fusion-protein corresponding to residues 1-258 of human cdc25C. Epitope mapped to residues 222-246.
Protein G chromatography
−20°C for 24 months.
Monoclonal