Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDC25C, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133687

Catalog No. NBP133687


Add to cart

Description

Description

CDC25C Polyclonal antibody specifically detects CDC25C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CDC25C
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
CDC25C
Recombinant protein encompassing a sequence within the center region of human CDC25C. The exact sequence is proprietary.
53 kDa
100 ul
Cell Cycle and Replication
Primary
995
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
CDC 25, CDC25, cell division cycle 25 homolog C (S. cerevisiae), cell division cycle 25 homolog C (S. pombe), cell division cycle 25C, Dual specificity phosphatase Cdc25C, EC 3.1.3.48, mitosis inducer CDC25, M-phase inducer phosphatase 3, phosphotyrosine phosphatase
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only