Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdc25C Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526000

Catalog No. PIPA526000

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CDC25C is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 and triggers entry into mitosis. It is also thought to suppress p53-induced growth arrest.
Specifications

Specifications

CDC25C
Polyclonal
PBS with 0.09% sodium azide
P30307, P48967
CDC25, PPP1R60, M-phase inducer phosphatase 3, cell division cycle 25C, dual specificity phosphatase CDC25C, mitosis inducer CDC25, phosphotyrosine phosphatase, protein phosphatase 1, regulatory subunit 60
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12532, 995
Western Blot
Unconjugated
CDC25C
Liquid
CDC25C
KLH conjugated synthetic peptide between 114-142 amino acids from the central region of human CDC25C
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit