Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC34 Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610249

Catalog No. BDB610249


Add to cart

Description

Description

The human homologue of the Saccharomyces cerevisiae CDC34 gene appears to be a highly conserved, single copy gene that is expressed in many cell lines. Thehuman CDC34 cDNA can functionally substitute for the yeast CDC34 gene and, therefore, is a mammalian homologue of a group of yeast genes (CDC34, CDC4, and CDC53) required for the late G1-S phase transition. In yeast, CDC34 is a member of the large family of ubiquitin-conjugating ligases (UBC) that are involved in linking ubiquitin polypeptides to target proteins. It is thought that the role of CDC34 in cell cycle progression is related to the ubiquitination of G1cyclins. In addition, p53 and other proteins involved in growth control such as Fos, Myc, and Mos are targeted for proteolysis by ubiquitin-dependent mechanisms. Absence of CDC34 causes yeast cell cycle arrest prior to DNA synthesis. Thus, it will be important to determine the role of CDC34 and the other proteins involved in the G1-S transition in mammalian cells. This will be a significant step toward understanding the regulation of the initiation of DNA synthesis, as well as the mechanism controlling the cell cycle transition following DNA damage.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

CDC34
17
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit