Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC5L Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612362

Catalog No. BDB612362


Add to cart

Description

Description

Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes and occurs in two distinct catalytic steps. The first step involves cleavage of the 5 ft. exon and the production of a lariat intermediate. In the second step, the 3 ft.-splice site is cleaved and the exons are fused with concomitant release of the intron lariat. The spliceosome contains multiple snRNPs and a number of non-snRNP splicing factors. CDC5L is a non-snRNP component of the splicesome that is homologous to the yeast Cdc5 protein. The sequence for CDC5L contgains a helix-turn-helix DNA binding domain (DBD), four nuclear localization signals, and a hydrophilic, proline-rich central region that is similar to the transcriptional activating domain in Myb family transcription factors. CDC5L can interact with the nuclear PP1 inhibitor NIPP-1 and the WD40 domain protein PLRG1. Both of these interactions are critical for pre-mRNA splicing and may also be important for G 2/M phase transition. In addition, CDC5L translocates from the cytoplasm to the nucleus in the presence of serum. Thus, CDC5L localization and splicesome-regulating activity may be controlled by mitogen-activated signal transduction pathways.

Bioimaging, Immunofluorescence, Western Blotting

Specifications

Specifications

CDC5L
21
Unconjugated
Affinity Purified
Rat CDC5L aa. 109-303
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit