Learn More
MilliporeSigma™ anti-Cdc6, Clone: DCS-180,
Manufacturer:  MilliporeSigma™ 05550
Catalog No. 05-550-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Cdc6
DCS-180
10mM PBS, pH 7.4 and 0.05% sodium azide
Purified
Murine
IgG1
100μg
Primary
Human, Murine
Immunoprecipitation, Western Blot
Unconjugated
NM_001254
CDC6, HsCDC18, cdc6, HsCDC6, p62, CDC18L
Recombinant, full-length human cdc6
Protein G chromatography
−20°C for 24 months.
Monoclonal