Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDCA8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511506

Catalog No. PIPA511506

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CDCA8 is a component of the chromosomal passenger complex (CPC) required for stability of the bipolar mitotic spindle. The CPC complex, a key regulator of mitosis, has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. In the complex, CDCA8 may be required to direct the CPC to centromeric DNA.
Specifications

Specifications

CDCA8
Polyclonal
PBS with 0.09% sodium azide
Q53HL2
Borealin; Dasra-B; hDasra-B; Cell division cycle-associated protein 8; Pluripotent embryonic stem cell-related gene 3 protein; CDCA8; PESCRG3;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55143
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CDCA8
Liquid
CDCA8
KLH conjugated synthetic peptide between 115-144 amino acids from the central region of human CDCA8
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit