Learn More
CDH20 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA526869
Catalog No. PIPA526869
Add to cart
Description
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
This gene is a type II classical cadherin from the cadherin superfamily and one of three cadherin 7-like genes located in a cluster on chromosome 18. The encoded membrane protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Since disturbance of intracellular adhesion is a prerequisite for invasion and metastasis of tumor cells, cadherins are considered prime candidates for tumor suppressor genes.
Specifications
Specifications
CDH20
Polyclonal
Unconjugated
CDH20
Liquid
CDH20
KLH conjugated synthetic peptide between 111-140 amino acids from the N-terminal region of human CDH20
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q9HBT6
CDH7L3, Cdh7, cadherin-20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
28316