Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cadherin 3, type 1, P-cadherin (placental), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CDH3 protein.

Manufacturer:  Abnova Corporation H00001001B01

Catalog No. 89-005-099


Add to cart

Description

Description

This gene is a classical cadherin from the cadherin superfamily. The encoded protein is a calcium-dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. This gene is located in a six-cadherin cluster in a region on the long arm of chromosome 16 that is involved in loss of heterozygosity events in breast and prostate cancer. In addition, aberrant expression of this protein is observed in cervical adenocarcinomas. Mutations in this gene have been associated with congential hypotrichosis with juvenile macular dystrophy. [provided by RefSeq

Sequence: MGLPRGPLASLLLLQVCWLQCAASEPCRAVFREAEVTLEAGGAEQEPGQALGKVFMGCPGQEPALFSTDNDDFTVRNGETVQERRSLKERNPLKIFPSKRILRRHKRDWVVAPISVPENGKGPFPQRLNQLKSNKDRDTKIFYSITGPGADSPPEGVFAVEKETGWLLLNKPLDREEIAKYELFGHAVSENGASVEDPMNISIIVTDQNDHKPKFTQDTFRGSVLEGVLPGTSVMQMTATDEDDAIYTYNGVVAYSIHSQEPKDPHDLMFTIHRSTGTISVISSGLDREKVPEYTLTIQATDMDGDGSTTTAVAVVEILDANDNAPMFDPQKYEAHVPENAVGHEVQRLTVTDLDAPNSPAWRATYLIMGGDDGDHFTITTHPESNQGILTTRKGLDFEAKNQHTLYVEVTNEAPFVLKLPTSTATIVVHVEDVNEAPVFVPPSKVVEVQEGIPTGEPVCVYTAEDPDKENQKISYRILRDPAGWLAMDPDSGQVTAVGTLDREDEQFVRNNIYEVMVLAMDNGSPPTTGTGTLLLTLIDVNDHGPVPEPRQITICNQSPVRQVLNITDKDLSPHTSPFQAQLTDDSDIYWTAEVNEEGDTVVLSLKKFLKQDTYDVHLSLSDHGNKEQLTVIRATVCDCHGHVETCPGPWKGGFILPVLGAVLALLFLLLVLLLLVRKKRKIKEPLLLPEDDTRDNVFYYGEEGGGEEDQDYDITQLHRGLEARPEVVLRNDVAPTIIPTPMYRPRPANPDEIGNFIIENLKAANTDPTAPPYDTLLVFDYEGSGSDAASLSSLTSSASDQDQDYDYLNEWGSRFKKLADMYGGGEDD
Specifications

Specifications

cadherin 3, type 1, P-cadherin (placental)
Unconjugated
No additive
BC041846.1
CDHP, HJMD, PCAD
Murine
50μL
Cell Adhesion
Y
Polyclonal
Human
Western Blot
cadherin 3, type 1, P-cadherin (placental)
CDH3
Serum
CDH3
CDH3 (AAH41846.1, 1 a.a. ∽829 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1001
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit