Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk1/Cdc2 (pY15) Mouse, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612307

 View more versions of this product

Catalog No. BDB612307


Add to cart

Description

Description

Progression of the mammalian cell cycle is regulated by phosphorylation of many key proteins. Several classes of cyclins (A-E) act as regulatory subunits for cyclin-dependent kinases (cdks). Cdk1/Cdc2 (p34 [cdc2]) is the catalytic subunit of the maturation promoting factor (MPF), which includes the regulatory subunit cyclin B. During late S and G2 phase, cyclin B synthesis increases, allowing it to bind Cdc2. This begins the transition into M-phase of the mammalian cell cycle by initiating a series of phosphorylation and dephosphorylation events that lead to activation of the Cdc2/cyclin B complex. After binding to cyclin B, cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex during G2 phase. Phosphorylation of cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the end of G2 phase and completes activation of the cdc2/Cyclin B complex and facilitates entry into mitosis. During mitosis, cyclin B is targeted for degradation and Cdc2 becomes inactive again.

Immunofluorescence, Western Blotting

Specifications

Specifications

Cdk1/Cdc2 (pY15)
44
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
p34 [cdc2]
Affinity Purified
Phosphorylated Human Cdc2 (pY15) Peptide
150μg
Cell Biology
Primary
Human, Murine, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.