Learn More
MilliporeSigma™ anti-Cdk1, Clone: A17.1.1, ™ (Chemicon™)
Supplier:  MilliporeSigma™ MAB8878/DEL
Encompass
Catalog No. 50-175-856
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative. View substitute products »
Description
Description
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Kinase Assay, Western Blotting
Specifications
Specifications
Cdk1
Unconjugated
Xenopus p34cdc2
RUO
Primary
Semi-purified
Monoclonal
Mouse
50 μg
Epigenetics
Chicken, Human, Mouse, Rat, Xenopus species
IgG2a
Recommendations
Recommendations