Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDK20, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132415

Catalog No. NBP132415


Add to cart

Description

Description

CDK20 Polyclonal antibody specifically detects CDK20 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CDK20
0.8mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
CDK20
Recombinant protein encompassing a sequence within the center region of human CCRK. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CAK-kinase p42, CCRKEC 2.7.11.22, CDCHP42, CDK-activating kinase p42, cell cycle related kinase, Cell cycle-related kinase, Cell division protein kinase 20, cyclin-dependent kinase 20, Cyclin-dependent protein kinase H, Cyclin-kinase-activating kinase p42, EC 2.7.11, p42, PNQALRE
Rabbit
IgG
0.1 ml
Protein Kinase
Primary
23552
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only