Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK4, Mouse, Clone: 3E7, Abnova™

Mouse monoclonal antibody raised against a partial recombinant CDK4.

Manufacturer:  Abnova Corporation H00001019M08

Catalog No. 89-021-025


  Check Availability
Add to cart

Specifications

Description

The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is highly similar to the gene products of S. cerevisiae cdc28 and S. pombe cdc2. It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression. The activity of this kinase is restricted to the G1-S phase, which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a). This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb). Mutations in this gene as well as in its related proteins including D-type cyclins, p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers. Multiple polyadenylation sites of this gene have been reported. [provided by RefSeq

Sequence: KPLFCGNSEADQLGKIFDLIGLPPEDDWPRDVSLPRGAFPPRGPRPVQSVVPEMEESGAQLLLEMLTFNPHKRISAFRALQHSYLHKDEGNPE
Description & Specifications

Specifications

CDK4
3E7
cyclin-dependent kinase 4
CDK4
Liquid
CDK4
CDK4 (AAH03644, 211 a.a. ∽304 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26kDa
100μg
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA,Western Blot
Unlabeled
In 1X PBS, pH 7.4
BC003644
CMM3, MGC14458, PSK-J3
Murine
IgG2b, κ
RUO
Primary
1019
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit