Learn More
CDK4 (M08A), Mouse anti-Human, Clone: 3E7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDK4.
Supplier: Abnova Corporation H00001019M08A
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is highly similar to the gene products of S. cerevisiae cdc28 and S. pombe cdc2. It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression. The activity of this kinase is restricted to the G1-S phase, which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a). This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb). Mutations in this gene as well as in its related proteins including D-type cyclins, p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers. Multiple polyadenylation sites of this gene have been reported. [provided by RefSeq
Sequence: KPLFCGNSEADQLGKIFDLIGLPPEDDWPRDVSLPRGAFPPRGPRPVQSVVPEMEESGAQLLLEMLTFNPHKRISAFRALQHSYLHKDEGNPESpecifications
CDK4 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC003644 | |
CDK4 | |
CDK4 (AAH03644, 211 a.a. ∼ 303 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
1019 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Western Blot | |
3E7 | |
Mouse monoclonal antibody raised against a partial recombinant CDK4. | |
CDK4 | |
CMM3/MGC14458/PSK-J3 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.