Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdk4 Mouse, Unlabeled, Clone: 97, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610148

 View more versions of this product

Catalog No. BDB610148


Add to cart

Description

Description

The Cdk4 kinase is a cell cycle-regulated cyclin-dependent kinase that associates with cyclins D1, D2, and D3. In macrophages and fibroblasts, Cdk4-Cyclin D1 complexes are predominant. Much like Cdk2 and Cdc2, the activity of Cdk4 is regulated by association with its respective kinase and by phosphorylation of specific threonine residues. The cdk-activating kinase (CAK) is a regulator of cdk-cyclin activity by virtue of its ability to phosphorylate single threonine residues on Cdk2, Cdc2, and Cdk4. Active Cdk4-Cyclin D1 complexes are capable of phosphorylating the retinoblastoma gene product (pRb), but not histone H1 or casein. Formation of the Cdk4-Cyclin D holoenzyme and phosphorylation of the catalytic subunit appear to be independently regulated. Therefore, other cellular factors may be necessary for stabilization of this protein complex in order to facilitate the phosphorylation of Cdk4. An inhibitor of Cdk4 kinase activity, known as p16, has been identified and partially characterized. The p16 gene appears to be altered in many tumors, suggesting that this is a tumor suppressor gene. It is thought that p16 inhibition of Cdk4-Cyclin D interferes with cell cycle progression by preventing phosphorylation of pRb by Cdk4 and the subsequent release of factors that activate transcription.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

Cdk4
97
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.