Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDK5RAP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526045

Catalog No. PIPA526045

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifically inhibits CDK5 activation by CDK5R1.
Specifications

Specifications

CDK5RAP1
Polyclonal
PBS with 0.09% sodium azide
Q96SZ6
C20orf34, C42, CDK5RAP1.3, CDK5RAP1.4, CGI-05, HSPC167, CDK5 activator-binding protein C42, CDK5 regulatory subunit-associated protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51654
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CDK5RAP1
Liquid
CDK5RAP1
KLH conjugated synthetic peptide between 544-571 amino acids from the C-terminal region of human CDK5RAP1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit