Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Cdk7, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215847
Description
Cdk7 Polyclonal specifically detects Cdk7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cdk7 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
CAK1Xenopus laevis, cdk-activating kinase), CDK-activating kinase, Cell division protein kinase 7, cyclin-dependent kinase 7, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, EC 2.7.11.24, p39 Mo15, serine/threonine kinase stk1,39 kDa protein kinase, serine/threonine protein kinase 1, serine/threonine protein kinase MO15, TFIIH basal transcription factor complex kinase subunit | |
Rabbit | |
39 kDa | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators | |
1022 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.04mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
P50613 | |
CDK7 | |
Recombinant protein encompassing a sequence within the center region of human CDK7. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Expected to cross react based on sequence identity: Xenopus laevis (86%), Zebrafish (84%), Bovine (96%), Xenopus tropicalis (87%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only