Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CDKAL1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136943

Catalog No. NBP136943


Add to cart

Description

Description

CDKAL1 Polyclonal antibody specifically detects CDKAL1 in Human, Mouse, Rat, Canine samples. It is validated for Peptide ELISA.
Specifications

Specifications

CDKAL1
0.5 mg/mL
Peptide ELISA Detection limit 1:64000
NP_060244.2
CDK5 regulatory subunit associated protein 1-like 1, CDK5 regulatory subunit-associated protein 1-like 1, FLJ20342, FLJ46705, MGC75469
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Canine, Human, Mouse, Rat
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
CDKAL1
Peptide with sequence C-QVKKQRTKDLSR corresponding to internal region according to NP_060244.2.
Immunogen affinity purified
RUO
Primary
54901
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only