Learn More
CDKL2 Mouse anti-Human, Clone: 6H8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA520569
Catalog No. PIMA520569
Add to cart
Description
Description
Peptide Sequence: LKDCSNVSVD HTRNPSVAIP PLTHNLSAVA PSINSGMGTE TIPIQGYRVD EKTKKCSIPF VKPNRHSPSG IYNINVTTLV SGPPLSDDSG ADLPQMEHQH
This gene product is a member of a large family of CDC2-related serine/threonine protein kinases. It accumulates primarily in the cytoplasm, with lower levels in the nucleus.
Specifications
Specifications
CDKL2
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q92772
Cyclin-dependent kinase-like 2, Serine/threonine-protein kinase KKIAMRE, Protein kinase p56 KKIAMRE, CDKL2,
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8999
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
6H8
Unconjugated
CDKL2
Liquid
CDKL2
CDKL2 (NP_003939, 394 a.a. ∽ 493 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human