Learn More
p57 Kip2 Rabbit anti-Bovine, Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA126615
Catalog No. PIPA126615
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-26615 detects cyclin-dependent kinase inhibitor 1C (p57, Kip2) from human, mouse, and bovine samples. PA1-26615 has been successfully used in immunohistochemistry (paraffin) procedures. The PA1-26615 immunogen is a synthetic peptide corresponding to a portion of human CDKN1C.
The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
Specifications
p57 Kip2
Polyclonal
PBS with sodium azide
P49918, P49919
p57 Kip2
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
1028, 12577, 510972
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
CDKN1C
Liquid
CDKN1C
Synthetic peptide corresponding to a portion of human CDKN1C.
IgG fraction
RUO
Antibody
Polyclonal
Bovine, Human, Mouse